ADV465750588 AChemBlock

4-Amino-1-[(2-chlorophenyl)methyl]pyrrolidin-2-one

  • MDL number MFCD12190575
  • Cas Number : 1114822-42-2
  • Molecular Weight : 224.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/4-amino-1-2-chlorophenyl-methyl-pyrrolidin-2-one.html

Safety Information

signalword Warning
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13678
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart