ADV465750593 AChemBlock

3-Ethenyloxetan-3-ol

  • MDL number MFCD14586454
  • Cas Number : 1207175-07-2
  • Molecular Weight : 100.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3-ethenyloxetan-3-ol.html

Safety Information

signalword warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13683
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart