ADV465750600 AChemBlock

5,7-Dichloropyrido[3,4-b]pyrazine

  • MDL number MFCD19707629
  • Cas Number : 1379338-74-5
  • Molecular Weight : 200.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/5-7-dichloropyrido-3-4-b-pyrazine.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13690
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart