ADV465750602 AChemBlock

Methyl 3-hydroxy-3-(trifluoromethyl)cyclobutane-1-carboxylate

  • MDL number MFCD23106188
  • Cas Number : 1392803-31-4
  • Molecular Weight : 198.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/methyl-3-hydroxy-3-trifluoromethyl-cyclobutane-1-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13692
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart