ADV465750605 AChemBlock

(1R,3S,5S)-3-methoxy-8-azabicyclo[3.2.1]octane hydrochloride

  • MDL number MFCD26959745
  • Cas Number : 1421254-66-1
  • Molecular Weight : 177.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1r-3s-5s-3-methoxy-8-azabicyclo-3-2-1-octane-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13695
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart