ADV465750610 AChemBlock

1-Amino-1H-pyrrole-2-carboxamide hydrochloride

  • MDL number MFCD28502519
  • Cas Number : 1630906-75-0
  • Molecular Weight : 161.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1-amino-1h-pyrrole-2-carboxamide-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13700
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart