ADV465750611 AChemBlock

2-(1-Methanesulfonylazetidin-3-ylidene)acetonitrile

  • MDL number MFCD28901359
  • Cas Number : 1638759-68-8
  • Molecular Weight : 172.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/2-1-methanesulfonylazetidin-3-ylidene-acetonitrile.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13701
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart