ADV465750613 AChemBlock

methyl (1R,2S)-2-((tert-butoxycarbonyl)amino)cyclopropane-1-carboxylate

  • MDL number MFCD28501607
  • Cas Number : 170299-60-2
  • Molecular Weight : 215.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/cis-cyclopropanecarboxylic-acid-2-1-1-dimethylethoxy-carbonyl-amino-methyl-ester.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13703
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart