ADV465750615 AChemBlock

3-Bromo-4-methoxy-1H-pyrazolo[3,4-d]pyrimidine

  • MDL number MFCD27995010
  • Cas Number : 190430-36-5
  • Molecular Weight : 229.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3-bromo-4-methoxy-1h-pyrazolo-3-4-d-pyrimidine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13705
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart