ADV465750627 AChemBlock

[(2S)-4,4-Difluoropyrrolidin-2-yl]methanol hydrochloride

  • MDL number MFCD22570251
  • Cas Number : 623583-10-8
  • Molecular Weight : 173.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form White powder
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/2s-4-4-difluoropyrrolidin-2-yl-methanol-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13717
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart