ADV465750628 AChemBlock

1-tert-Butyl 2-methyl (2R)-4,4-difluoropyrrolidine-1,2-dicarboxylate

  • MDL number MFCD11976647
  • Cas Number : 647857-74-7
  • Molecular Weight : 265.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1-tert-butyl-2-methyl-2r-4-4-difluoropyrrolidine-1-2-dicarboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13718
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart