ADV465750634 AChemBlock

3-Benzyl-6-oxa-3-azabicyclo[3.1.0]hexane

  • MDL number MFCD09608024
  • Cas Number : 75390-09-9
  • Molecular Weight : 175.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3-benzyl-6-oxa-3-azabicyclo-3-1-0-hexane.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13724
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart