ADV465750646 AChemBlock

1-{5-Bromo-1H-pyrrolo[2,3-b]pyridin-3-yl}ethan-1-one

  • MDL number MFCD12403936
  • Cas Number : 866545-96-2
  • Molecular Weight : 239.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1-5-bromo-1h-pyrrolo-2-3-b-pyridin-3-yl-ethan-1-one.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13737
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart