ADV465750667 AChemBlock

tert-butyl 3-oxo-2,7-diazaspiro[4.5]decane-7-carboxylate

  • MDL number MFCD11227061
  • Cas Number : 1158750-91-4
  • Molecular Weight : 254.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/2-7-diazaspiro-4-5-decane-7-carboxylic-acid-3-oxo-1-1-dimethylethyl-ester.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13758
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart