ADV465750671 AChemBlock

Methyl 3-bromoimidazo[1,5-a]pyridine-1-carboxylate

  • MDL number MFCD22566113
  • Cas Number : 1039357-00-0
  • Molecular Weight : 255.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/methyl-3-bromoimidazo-1-5-a-pyridine-1-carboxylate.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13762
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart