ADV465750672 AChemBlock

3-Bromo-1-(oxan-2-yl)-1H-pyrazole

  • MDL number MFCD16659986
  • Cas Number : 1044270-96-3
  • Molecular Weight : 231.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3-bromo-1-oxan-2-yl-1h-pyrazole.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13763
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart