ADV465750678 AChemBlock

5H-Pyrrolo[3,2-d]pyrimidin-2-amine

  • MDL number MFCD09800376
  • Cas Number : 943736-60-5
  • Molecular Weight : 134.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form Light yellow solid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/5h-pyrrolo-3-2-d-pyrimidin-2-amine.html

Safety Information

signalword Warning
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13158
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart