ADV465750679 AChemBlock

4-Hydroxy-1,2,5,6,7,8-hexahydroquinolin-2-one

  • MDL number MFCD13179322
  • Cas Number : 56517-59-0
  • Molecular Weight : 165.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/4-hydroxy-1-2-5-6-7-8-hexahydroquinolin-2-one.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13770
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart