ADV465750683 AChemBlock

(8aS)-Hexahydro-1H-[1,3]oxazolo[3,4-a]piperazin-3-one hydrochloride

  • MDL number MFCD09878826
  • Cas Number : 958635-15-9
  • Molecular Weight : 178.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/8as-hexahydro-1h-1-3-oxazolo-3-4-a-piperazin-3-one-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13774
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart