ADV465750709 AChemBlock

Imidazo[1,2-a]pyridin-7-amine hydrobromide

  • MDL number MFCD28501213
  • Cas Number : 1630906-80-7
  • Molecular Weight : 214.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/imidazo-1-2-a-pyridin-7-amine-hydrobromide.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13800
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart