ADV465750712 AChemBlock

3-Hydroxycyclobutane-1-carbonitrile

  • MDL number MFCD13659429
  • Cas Number : 20249-17-6
  • Molecular Weight : 97.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form Colorless liquid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3-hydroxycyclobutane-1-carbonitrile.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13803
pictograms GHS02
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart