ADV465750720 AChemBlock

N-Methoxy-N-methylcyclohexanecarboxamide

  • MDL number MFCD12406909
  • Cas Number : 80783-98-8
  • Molecular Weight : 171.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/n-methoxy-n-methylcyclohexanecarboxamide.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13811
pictograms GHS05
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart