ADV465750752 AChemBlock

1- (Phenysulfonyl)-2-iodo-5-methoxy-7-azaindole

  • MDL number MFCD16875774
  • Cas Number : 1227267-13-1
  • Molecular Weight : 414.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1-phenysulfonyl-2-iodo-5-methoxy-7-azaindole.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=M15024
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart