ADV465750762 AChemBlock

6-Chloro-5-methoxy-1H-indazole

  • MDL number MFCD20923428
  • Cas Number : 13096-98-5
  • Molecular Weight : 182.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/6-chloro-5-methoxy-1h-indazole.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=M15034
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart