ADV465750777 AChemBlock

4-Fluoro-1H-indole-5-carboxylic acid

  • MDL number MFCD12924428
  • Cas Number : 908600-70-4
  • Molecular Weight : 179.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/4-fluoro-1h-indole-5-carboxylic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=M15052
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart