ADV465750787 AChemBlock

Methyl 4-chloro-1H-indazole-6-carboxylate

  • MDL number MFCD07781359
  • Cas Number : 885519-19-7
  • Molecular Weight : 210.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/methyl-4-chloro-1h-indazole-6-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=M15062
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart