ADV465750793 AChemBlock

Ethyl 4-fluoro-1H-indole-7-carboxylate

  • MDL number MFCD26143075
  • Cas Number : 1196048-19-7
  • Molecular Weight : 207.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/ethyl-4-fluoro-1h-indole-7-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=M15068
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart