ADV465750798 AChemBlock

2-Trifluoromethyl-5-iodo-4-amino-pyridine

  • MDL number MFCD28672666
  • Cas Number : 1239462-10-2
  • Molecular Weight : 288.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/2-trifluoromethyl-5-iodo-4-amino-pyridine.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=M15073
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart