ADV465750800 AChemBlock

Methyl 5-bromo-6-methyl-7-azaindole-3-carboxylate

  • MDL number MFCD21606703
  • Cas Number : 1881328-63-7
  • Molecular Weight : 269.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/methyl-5-bromo-6-methyl-7-azaindole-3-carboxylate.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=M15075
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart