ADV465750819 AChemBlock

4-Iodo-1,3-dihydro-2H-indol-2-one

  • MDL number MFCD09835632
  • Cas Number : 179536-52-8
  • Molecular Weight : 259.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/4-iodo-1-3-dihydro-2h-indol-2-one.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=M15095
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart