ADV465750820 AChemBlock

3-Hydroxy 1H-indazole-5-methylcarboxylate

  • MDL number MFCD11007794
  • Cas Number : 1082041-20-0
  • Molecular Weight : 192.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3-hydroxy-1h-indazole-5-methylcarboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=M15096
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart