ADV465750823 AChemBlock

4-Bromo-1H-indazole-3-carbonitrile

  • MDL number MFCD11845593
  • Cas Number : 1167055-55-1
  • Molecular Weight : 222.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/4-bromo-1h-indazole-3-carbonitrile.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=M15099
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart