ADV465750832 AChemBlock

3-Bromo-4-nitro-1H-indazole

  • MDL number MFCD07781572
  • Cas Number : 74209-17-9
  • Molecular Weight : 242.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3-bromo-4-nitro-1h-indazole.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=M15108
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart