ADV465750836 AChemBlock

Methyl 4-(benzyloxy)-1H-indole-6-carboxylate

  • MDL number MFCD11110567
  • Cas Number : 61545-36-6
  • Molecular Weight : 281.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/methyl-4-benzyloxy-1h-indole-6-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=M15112
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart