ADV465750838 AChemBlock

3-(Methoxycarbonyl)-4-nitrobenzoic acid

  • MDL number MFCD09026978
  • Cas Number : 64152-09-6
  • Molecular Weight : 225.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3-methoxycarbonyl-4-nitrobenzoic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=M15114
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart