ADV465750879 AChemBlock

ABT 578

  • MDL number MFCD09752954
  • Cas Number : 221877-54-9
  • Molecular Weight : 966.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/abt-578.html

Safety Information

signalword warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=R16067
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart