ADV465750920 AChemBlock

(3S)-1-[(2,4-dimethoxyphenyl)methyl]-5-oxopyrrolidine-3-carboxylic acid

  • MDL number MFCD28001381
  • Cas Number : 755025-16-2
  • Molecular Weight : 279.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3s-1-2-4-dimethoxyphenyl-methyl-5-oxopyrrolidine-3-carboxylic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9927
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart