ADV509447589 AChemBlock

(3S)-3-Hydroxy-D-proline

  • MDL number MFCD09996919
  • Cas Number : 118492-86-7
  • Molecular Weight : 131.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3s-3-hydroxy-d-proline.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4332
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart