ADV509447595 AChemBlock

1-(Hydroxymethyl)-2-azabicyclo[2.2.2]octan-3-one

  • MDL number MFCD20261415
  • Cas Number : 1334412-35-9
  • Molecular Weight : 155.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1-hydroxymethyl-2-azabicyclo-2-2-2-octan-3-one.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=H-8237
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart