ADV509447603 AChemBlock

1-(5-bromo-1,3-thiazol-2-yl)ethan-1-one

  • MDL number MFCD09842273
  • Cas Number : 774230-98-7
  • Molecular Weight : 206.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/5-bromothiazole-2-ethanone.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9633
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart