ADV509447615 AChemBlock

3-Fluorocyclobutan-1-one

  • MDL number MFCD23106394
  • Cas Number : 1415342-84-5
  • Molecular Weight : 88.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3-fluorocyclobutan-1-one.html

Safety Information

signalword Warning
pcodes P210;P233;P280;P303+361+351
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=H-8760
pictograms GHS02
hstatements H226
Availability details for
Quantity
  • Availability
  • Add to Cart