ADV509447617 AChemBlock

Ethyl 1-Boc-amino-3-oxocyclobutane-1-carboxylate

  • MDL number MFCD18633152
  • Cas Number : 129287-91-8
  • Molecular Weight : 257.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/ethyl-1-boc-amino-3-oxocyclobutane-1-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=H-8781
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart