ADV509447628 AChemBlock

Endo-8-azabicyclo[3.2.1]octane-3-methanol hydrochloride

  • MDL number MFCD23381625
  • Cas Number : 1257442-93-5
  • Molecular Weight : 177.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/endo-8-azabicyclo-3-2-1-octane-3-methanol-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13207
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart