ADV509447631 AChemBlock

(1R,3R)-3-Aminocyclopentan-1-ol hydrochloride

  • MDL number MFCD12405849
  • Cas Number : 1523541-74-3
  • Molecular Weight : 137.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1r-3r-3-aminocyclopentan-1-ol-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13346
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart