ADV509447634 AChemBlock

Methyl 5-bromo-1H-pyrrolo[2,3-b]pyridine-3-carboxylate

  • MDL number MFCD15529254
  • Cas Number : 872619-43-7
  • Molecular Weight : 255.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/methyl-5-bromo-1h-pyrrolo-2-3-b-pyridine-3-carboxylate.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13730
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart