ADV509447649 AChemBlock

7-Bromo-2H-pyrido[3,2-b][1,4]oxazin-3(4H)-one

  • MDL number MFCD09834134
  • Cas Number : 122450-96-8
  • Molecular Weight : 229.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/7-bromo-2h-pyrido-3-2-b-1-4-oxazin-3-4h-one.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=H-8933
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart