ADV509447654 AChemBlock

tert-Butyl hexahydro-1H-pyrrolo[3,4-c]pyridine-5(6H)-carboxylate hydrochloride

  • MDL number MFCD12913817
  • Cas Number : 1187933-06-7
  • Molecular Weight : 262.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-hexahydro-1h-pyrrolo-3-4-c-pyridine-5-6h-carboxylate-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=H-8940
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart