ADV509447676 AChemBlock

LY-3009120

  • MDL number MFCD28411374
  • Cas Number : 1454682-72-4
  • Molecular Weight : 424.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/ly-3009120.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=H-8969
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart