ADV509447707 AChemBlock

Methyl 2-(4-methylpiperidin-3-yl)acetate hydrochloride

  • MDL number MFCD30146429
  • Cas Number : 1860028-39-2
  • Molecular Weight : 207.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/methyl-2-4-methylpiperidin-3-yl-acetate-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13831
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart