ADV509447709 AChemBlock

3-Bromo-7-iodothieno[3,2-c]pyridin-4-amine

  • MDL number MFCD14706353
  • Cas Number : 799293-91-7
  • Molecular Weight : 355

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-bromo-7-iodothieno-3-2-c-pyridin-4-amine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13833
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart